ENA370562900 Enamine

2-methoxy-5-phenylbenzaldehyde

  • MDL number MFCD04039099
  • Cas Number : 89536-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104305

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104305
Availability details for
Quantity
  • Availability
  • Add to Cart