ENA370562901 Enamine

4-chloro-6-methoxypyrimidin-5-amine

  • MDL number MFCD01646119
  • Cas Number : 15846-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104306

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104306
Availability details for
Quantity
  • Availability
  • Add to Cart