ENA370562906 Enamine

2-[(4-bromophenyl)methyl]-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD14648368
  • Cas Number : 950785-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-104311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104311
Availability details for
Quantity
  • Availability
  • Add to Cart