ENA370562911 Enamine

[4-methyl-2-(pyrrolidin-1-yl)phenyl]methanamine

  • MDL number MFCD16704798
  • Cas Number : 1248232-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104361

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104361
Availability details for
Quantity
  • Availability
  • Add to Cart