ENA370562916 Enamine

2-(2-bromophenyl)pyrrolidine hydrochloride

  • MDL number MFCD08456883
  • Cas Number : 1197232-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104371

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104371
Availability details for
Quantity
  • Availability
  • Add to Cart