ENA370562917 Enamine

[2-(2-ethoxyethoxy)-4-methylphenyl]methanamine

  • MDL number MFCD16704886
  • Cas Number : 1248259-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104377

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104377
Availability details for
Quantity
  • Availability
  • Add to Cart