ENA370562918 Enamine

5-(4-bromophenyl)-1H-imidazol-2-amine

  • MDL number MFCD03308789
  • Cas Number : 60472-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104378

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-104378
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart