ENA370562924 Enamine

1-{4-[4-(aminomethyl)phenyl]piperazin-1-yl}ethan-1-one

  • MDL number MFCD11135267
  • Cas Number : 1019625-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101897

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101897
Availability details for
Quantity
  • Availability
  • Add to Cart