ENA370562925 Enamine

2,2-dimethylcyclohexan-1-amine

  • MDL number MFCD16658197
  • Cas Number : 24247-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101901

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-101901
pictograms GHS02,GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart