ENA370562926 Enamine

cyclopentyl(2-fluorophenyl)methanamine

  • MDL number MFCD16303686
  • Cas Number : 1226407-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104388

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104388
Availability details for
Quantity
  • Availability
  • Add to Cart