ENA370562931 Enamine

methyl 2-amino-6-chloro-3-methylbenzoate

  • MDL number MFCD12159916
  • Cas Number : 298221-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104404

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104404
Availability details for
Quantity
  • Availability
  • Add to Cart