ENA370562933 Enamine

2-[(3-methoxyphenyl)amino]acetic acid

  • MDL number MFCD06849831
  • Cas Number : 85676-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102049
Availability details for
Quantity
  • Availability
  • Add to Cart