ENA370562939 Enamine

4-bromo-2-nitro-1-phenoxybenzene

  • MDL number MFCD13191935
  • Cas Number : 56966-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102185

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-102185
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart