ENA370562944 Enamine

(5-chloro-1,3,4-thiadiazol-2-yl)methanol

  • MDL number MFCD14155682
  • Cas Number : 912669-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102287
Availability details for
Quantity
  • Availability
  • Add to Cart