ENA370562957 Enamine

methyl 2-(tert-butylsulfanyl)acetate

  • MDL number MFCD11173895
  • Cas Number : 49827-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104475
Availability details for
Quantity
  • Availability
  • Add to Cart