ENA370562960 Enamine

methyl 2-aminocycloheptane-1-carboxylate hydrochloride

  • MDL number MFCD09910329
  • Cas Number : 1198283-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-148
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104478
Availability details for
Quantity
  • Availability
  • Add to Cart