ENA370562964 Enamine

4-amino-1,1-dioxo-1lambda6-thiane-4-carboxylic acid

  • MDL number MFCD09607995
  • Cas Number : 39124-27-1
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-aminotetrahydro-2H-thiopyran-4-carboxylic acid 1,1-dioxide
Inchi Code 1S/C6H11NO4S/c7-6(5(8)9)1-3-12(10,11)4-2-6/h1-4,7H2,(H,8,9)
Inchi Key FANFYQGRZWKLBT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104482

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-104482
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart