ENA370562967 Enamine

5-chloro-2-[4-(chloromethyl)phenyl]-1,3-benzothiazole

  • MDL number MFCD11534816
  • Cas Number : 1096267-76-3
  • Molecular Weight : 294.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104499
Availability details for
Quantity
  • Availability
  • Add to Cart