ENA370562968 Enamine

2-methoxyquinazolin-4-ol

  • MDL number MFCD01464100
  • Cas Number : 1011-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102428
Availability details for
Quantity
  • Availability
  • Add to Cart