ENA370562969 Enamine

(2-bromo-3-phenylpropyl)benzene

  • MDL number MFCD00666050
  • Cas Number : 89036-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104501

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104501
Availability details for
Quantity
  • Availability
  • Add to Cart