ENA370562971 Enamine

7-methyl-[1,2,4]triazolo[4,3-a]pyridin-3-amine

  • MDL number MFCD11847743
  • Cas Number : 5006-56-4
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 298-300
Iupac Name 7-methyl[1,2,4]triazolo[4,3-a]pyridin-3-ylamine
Inchi Code 1S/C7H8N4/c1-5-2-3-11-6(4-5)9-10-7(11)8/h2-4H,1H3,(H2,8,10)
Inchi Key BOMGURPAEXSYAI-UHFFFAOYSA-N

Description

Melting Point 298-300
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104505

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-104505
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart