ENA370562977 Enamine

1-(3-fluoro-4-methylphenyl)ethan-1-amine

  • MDL number MFCD09041163
  • Cas Number : 926262-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104514

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104514
Availability details for
Quantity
  • Availability
  • Add to Cart