ENA370562984 Enamine

(2-phenylpyrimidin-4-yl)methanol

  • MDL number MFCD16866629
  • Cas Number : 38705-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43...45
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104549

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104549
Availability details for
Quantity
  • Availability
  • Add to Cart