ENA370562993 Enamine

1-methyl-2,3-dihydro-1H-indol-4-amine

  • MDL number MFCD19207651
  • Cas Number : 801316-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104559

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104559
Availability details for
Quantity
  • Availability
  • Add to Cart