ENA370562994 Enamine

5,6-difluoro-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD16845540
  • Cas Number : 142356-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104574
Availability details for
Quantity
  • Availability
  • Add to Cart