ENA370562997 Enamine

7-methyl-1H,2H,3H,4H-pyrido[2,3-b]pyrazine

  • MDL number MFCD18252983
  • Cas Number : 1378818-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104577
Availability details for
Quantity
  • Availability
  • Add to Cart