ENA370563009 Enamine

[2-(hexyloxy)-4-methylphenyl]methanamine

  • MDL number MFCD16731732
  • Cas Number : 1247738-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-104699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104699
Availability details for
Quantity
  • Availability
  • Add to Cart