ENA370563023 Enamine

3-(4-bromophenyl)-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD17259821
  • Cas Number : 1341567-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104780

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104780
Availability details for
Quantity
  • Availability
  • Add to Cart