ENA370563024 Enamine

3-(1-benzofuran-2-yl)-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD17292416
  • Cas Number : 1343081-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104784

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104784
Availability details for
Quantity
  • Availability
  • Add to Cart