ENA370563041 Enamine

2-fluoro-4-methylbenzamide

  • MDL number MFCD06660328
  • Cas Number : 886761-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104869
Availability details for
Quantity
  • Availability
  • Add to Cart