ENA370563046 Enamine

4-chloronaphthalene-1-sulfonyl chloride

  • MDL number MFCD01365822
  • Cas Number : 64318-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-104874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104874
Availability details for
Quantity
  • Availability
  • Add to Cart