ENA370563051 Enamine

5-(2,5-dichlorophenyl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD18866980
  • Cas Number : 1339337-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104882
Availability details for
Quantity
  • Availability
  • Add to Cart