ENA370563052 Enamine

3-(4-bromobenzenesulfonyl)-2-methylpropanoic acid

  • MDL number MFCD10024398
  • Cas Number : 1017674-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104885
Availability details for
Quantity
  • Availability
  • Add to Cart