ENA370563053 Enamine

2-[(4-chlorobenzene)sulfonamido]-3-hydroxypropanoic acid

  • MDL number MFCD02230141

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104888

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104888
Availability details for
Quantity
  • Availability
  • Add to Cart