ENA370563054 Enamine

methyl 4-(2-amino-1,3-thiazol-4-yl)butanoate

  • MDL number MFCD22369870
  • Cas Number : 193817-77-5
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(2-amino-1,3-thiazol-4-yl)butanoate
Inchi Code 1S/C8H12N2O2S/c1-12-7(11)4-2-3-6-5-13-8(9)10-6/h5H,2-4H2,1H3,(H2,9,10)
Inchi Key YSWFMWBCWKBGMX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104889
Availability details for
Quantity
  • Availability
  • Add to Cart