ENA370563057 Enamine

2-[N-cyclopentyl-3-(1H-1,2,4-triazol-1-yl)propanamido]acetic acid

  • MDL number MFCD12525239
  • Cas Number : 1179749-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104894
Availability details for
Quantity
  • Availability
  • Add to Cart