ENA370563058 Enamine

(3-ethylthiophen-2-yl)methanamine

  • MDL number MFCD14656349
  • Cas Number : 1250601-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104897

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104897
Availability details for
Quantity
  • Availability
  • Add to Cart