ENA370563061 Enamine

methyl 2-(pyrazin-2-yl)propanoate

  • MDL number MFCD19622586
  • Cas Number : 1341956-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104901
Availability details for
Quantity
  • Availability
  • Add to Cart