ENA370563066 Enamine

1-[4-(4-chlorophenyl)-5-methyl-1,3-thiazol-2-yl]ethan-1-amine

  • MDL number MFCD05221598
  • Cas Number : 642929-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104906

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104906
Availability details for
Quantity
  • Availability
  • Add to Cart