ENA370563075 Enamine

(2,6-dimethoxyphenyl)methanol

  • MDL number MFCD00016862
  • Cas Number : 16700-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104986

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104986
Availability details for
Quantity
  • Availability
  • Add to Cart