ENA370563076 Enamine

2,4-dichloro-5-methanesulfonylbenzoic acid

  • MDL number MFCD11179857
  • Cas Number : 51521-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104988

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104988
Availability details for
Quantity
  • Availability
  • Add to Cart