ENA370563085 Enamine

2,6-difluoro-3-methanesulfonylbenzoic acid

  • MDL number MFCD11179943
  • Cas Number : 845617-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100728

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100728
Availability details for
Quantity
  • Availability
  • Add to Cart