ENA370563089 Enamine

2-chloro-3-hydrazinylpyridine

  • MDL number MFCD07786314
  • Cas Number : 117087-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-102824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102824
Availability details for
Quantity
  • Availability
  • Add to Cart