ENA370563103 Enamine

2-(propan-2-yl)-2H-1,2,3,4-tetrazol-5-amine

  • MDL number MFCD07432847
  • Cas Number : 229003-16-1
  • Molecular Weight : 127.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-isopropyl-2H-tetraazol-5-amine
Inchi Code 1S/C4H9N5/c1-3(2)9-7-4(5)6-8-9/h3H,1-2H3,(H2,5,7)
Inchi Key NEJVMYROZIQXMS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-105087

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-105087
pictograms GHS05,GHS07
hstatements H302,H312,H314,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart