ENA370563104 Enamine

methyl 4-methylpyrrolidine-3-carboxylate

  • MDL number MFCD03788462
  • Cas Number : 885952-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105092

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105092
Availability details for
Quantity
  • Availability
  • Add to Cart