ENA370563114 Enamine

N-ethyl-5-phenyl-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD12170507
  • Cas Number : 1155607-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105117
Availability details for
Quantity
  • Availability
  • Add to Cart