ENA370563116 Enamine

2-(2-fluorophenyl)-2-(phenylamino)acetic acid

  • MDL number MFCD12899046
  • Cas Number : 1146747-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105122

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105122
Availability details for
Quantity
  • Availability
  • Add to Cart