ENA370563122 Enamine

N-benzylpiperidin-3-amine

  • MDL number MFCD16066379
  • Cas Number : 396730-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105128

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-105128
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart