ENA370563126 Enamine

2-methyl-4-(2-methylpropyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD16304730
  • Cas Number : 1226165-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100814
Availability details for
Quantity
  • Availability
  • Add to Cart