ENA370563128 Enamine

(1S)-1-(furan-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD12910711

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103054
Availability details for
Quantity
  • Availability
  • Add to Cart